Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGCAGCTGGGGCTTAGGCGGGCC[A/G]CCCAGAAGCACAGCTTTCCTCGAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613317 MIM: 162080 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCAF11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCAF11 - DDB1 and CUL4 associated factor 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163484.1 | 1106 | Missense Mutation | ACC,GCC | T,A 126 | NP_001156956.1 | |
NM_025230.4 | 1106 | Missense Mutation | ACC,GCC | T,A 126 | NP_079506.3 | |
NM_181357.2 | 1106 | Missense Mutation | ACC,GCC | T,A 100 | NP_852002.1 |
NRL - neural retina leucine zipper | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |