Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCCTACCCTCCCTCCTCCTGGTA[T/C]GTGGCTGTGAGGTGCAGGCGGCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
36 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM213B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM213B - family with sequence similarity 213 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195736.1 | Intron | NP_001182665.1 | ||||
NM_001195737.1 | Intron | NP_001182666.1 | ||||
NM_001195738.1 | Intron | NP_001182667.1 | ||||
NM_001195740.1 | Intron | NP_001182669.1 | ||||
NM_001195741.1 | Intron | NP_001182670.1 | ||||
NM_152371.3 | Intron | NP_689584.2 | ||||
XM_006710354.3 | Intron | XP_006710417.1 | ||||
XM_011540664.1 | Intron | XP_011538966.1 | ||||
XM_011540665.1 | Intron | XP_011538967.1 | ||||
XM_011540666.1 | Intron | XP_011538968.1 |
LOC100996583 - uncharacterized LOC100996583 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MMEL1 - membrane metallo-endopeptidase-like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |