Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGCAGAATTAGACCCATTAGACA[C/T]GTATATTTCAGTTGTAATTTCAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604653 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC40A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC40A1 - solute carrier family 40 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014585.5 | 1780 | Missense Mutation | NP_055400.1 | |||
XM_005246505.1 | 1780 | Missense Mutation | XP_005246562.1 | |||
XM_017003938.1 | 1780 | Missense Mutation | XP_016859427.1 |