Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGAGAAGCCCACGGGCGTGAACGC[A/G]GCGGAAGTTCGCGAGATGGTCGCGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606377 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DHDH PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DHDH - dihydrodiol dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014475.3 | 909 | Silent Mutation | GCA,GCG | A,A 103 | NP_055290.1 | |
XM_005258748.4 | 909 | Intron | XP_005258805.1 | |||
XM_017026598.1 | 909 | Silent Mutation | GCA,GCG | A,A 20 | XP_016882087.1 |
Set Membership: |
HapMap |