Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATTTGGTCCTTACCCTTAACGTG[G/T]CTCTAAGACCAGAAGGGAACACCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616967 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA0753 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIAA0753 - KIAA0753 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014804.2 | Intron | NP_055619.2 | ||||
XM_006721612.2 | Intron | XP_006721675.1 | ||||
XM_011524090.2 | Intron | XP_011522392.1 | ||||
XM_011524091.2 | Intron | XP_011522393.1 | ||||
XM_011524095.2 | Intron | XP_011522397.1 | ||||
XM_011524096.1 | Intron | XP_011522398.1 | ||||
XM_011524098.2 | Intron | XP_011522400.1 | ||||
XM_011524099.2 | Intron | XP_011522401.1 | ||||
XM_011524100.2 | Intron | XP_011522402.1 | ||||
XM_017025455.1 | Intron | XP_016880944.1 | ||||
XM_017025456.1 | Intron | XP_016880945.1 | ||||
XM_017025457.1 | Intron | XP_016880946.1 |
MED31 - mediator complex subunit 31 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TXNDC17 - thioredoxin domain containing 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032731.3 | Intron | NP_116120.1 |