Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGGGTGATTTCCAAGGGCATCTGG[A/T]CACCAAGACAATACCAAAAAAAGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C2orf42 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C2orf42 - chromosome 2 open reading frame 42 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017880.1 | Intron | NP_060350.1 | ||||
XM_005264389.2 | Intron | XP_005264446.1 | ||||
XM_005264390.2 | Intron | XP_005264447.1 | ||||
XM_005264391.2 | Intron | XP_005264448.1 | ||||
XM_011532937.2 | Intron | XP_011531239.1 | ||||
XM_017004398.1 | Intron | XP_016859887.1 | ||||
XM_017004399.1 | Intron | XP_016859888.1 | ||||
XM_017004400.1 | Intron | XP_016859889.1 |