Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTTAATTTTTCTTAGAACTTAAC[A/G]CTGCTAGATCTAAAATATAAAAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610730 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCT6B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCT6B - chaperonin containing TCP1 subunit 6B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193529.2 | Intron | NP_001180458.1 | ||||
NM_001193530.1 | Intron | NP_001180459.1 | ||||
NM_006584.3 | Intron | NP_006575.2 | ||||
XM_011524203.2 | Intron | XP_011522505.1 | ||||
XM_017024023.1 | Intron | XP_016879512.1 | ||||
XM_017024024.1 | Intron | XP_016879513.1 |