Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCTCCAAGCTCCTGCTAGGCTCA[C/T]GCCTGTCTATTGCAGGGGCCCAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 100720 MIM: 100730 MIM: 612972 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHRND PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHRND - cholinergic receptor nicotinic delta subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CHRNG - cholinergic receptor nicotinic gamma subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005199.4 | Intron | NP_005190.4 |
TIGD1 - tigger transposable element derived 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |