Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCTGCCTGGGGCACAGCCTGGTA[C/T]AGATTCAGAGCCCTGCCCTTCCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120270 MIM: 611157 MIM: 606459 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL9A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COL9A3 - collagen type IX alpha 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRGBP - MRG/MORF4L binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OGFR - opioid growth factor receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007346.2 | Intron | NP_031372.2 |
OGFR-AS1 - OGFR antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |