Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCAGCCTGGGGGACCTCGGGCCT[C/T]TTCCGCGGGAGGTAGGGCGCGGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C6orf141 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C6orf141 - chromosome 6 open reading frame 141 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145652.1 | 243 | Missense Mutation | CTT,TTT | L,F 32 | NP_001139124.1 | |
XM_005248850.3 | 243 | Missense Mutation | CTT,TTT | L,F 32 | XP_005248907.1 | |
XM_005248852.2 | 243 | Missense Mutation | CTT,TTT | L,F 32 | XP_005248909.1 | |
XM_005248853.2 | 243 | Missense Mutation | CTT,TTT | L,F 32 | XP_005248910.1 | |
XM_006714997.2 | 243 | Missense Mutation | CTT,TTT | L,F 32 | XP_006715060.1 | |
XM_011514303.2 | 243 | Missense Mutation | CTT,TTT | L,F 32 | XP_011512605.1 | |
XM_017010294.1 | 243 | Missense Mutation | CTT,TTT | L,F 32 | XP_016865783.1 | |
XM_017010295.1 | 243 | Missense Mutation | CTT,TTT | L,F 32 | XP_016865784.1 | |
XM_017010296.1 | 243 | Missense Mutation | CTT,TTT | L,F 32 | XP_016865785.1 | |
XM_017010297.1 | 243 | Missense Mutation | CTT,TTT | L,F 32 | XP_016865786.1 | |
XM_017010298.1 | 243 | Missense Mutation | CTT,TTT | L,F 32 | XP_016865787.1 |