Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTCCTGAAGGAGCCCTCAATTGC[A/G]TTGGAAAAGCTGCGGGAACTCAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616218 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBC1D13 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TBC1D13 - TBC1 domain family member 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286772.1 | 157 | Silent Mutation | GCA,GCG | A,A 22 | NP_001273701.1 | |
NM_018201.4 | 157 | Silent Mutation | GCA,GCG | A,A 22 | NP_060671.3 | |
XM_005252060.2 | 157 | UTR 5 | XP_005252117.1 |