Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610429 MIM: 615967 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C7orf50 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C7orf50 - chromosome 7 open reading frame 50 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134395.1 | 179 | Intron | NP_001127867.1 | |||
NM_001134396.1 | 179 | Intron | NP_001127868.1 | |||
NM_001318252.1 | 179 | Intron | NP_001305181.1 | |||
NM_032350.5 | 179 | Intron | NP_115726.1 | |||
XM_006715793.2 | 179 | Intron | XP_006715856.1 | |||
XM_011515580.2 | 179 | Intron | XP_011513882.1 | |||
XM_011515581.2 | 179 | Intron | XP_011513883.1 | |||
XM_011515582.2 | 179 | Intron | XP_011513884.1 | |||
XM_011515583.2 | 179 | Intron | XP_011513885.1 | |||
XM_011515584.2 | 179 | Intron | XP_011513886.1 | |||
XM_017012719.1 | 179 | Intron | XP_016868208.1 | |||
XM_017012720.1 | 179 | Intron | XP_016868209.1 | |||
XM_017012721.1 | 179 | Intron | XP_016868210.1 | |||
XM_017012722.1 | 179 | Intron | XP_016868211.1 |
COX19 - COX19, cytochrome c oxidase assembly factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CYP2W1 - cytochrome P450 family 2 subfamily W member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017781.2 | 179 | Silent Mutation | CTG,TTG | L,L 56 | NP_060251.2 | |
XM_011515440.2 | 179 | Silent Mutation | CTG,TTG | L,L 56 | XP_011513742.1 | |
XM_011515441.2 | 179 | Silent Mutation | CTG,TTG | L,L 56 | XP_011513743.1 |
Set Membership: |
Validated |