Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGCGGTCTGGGACTCCTCCTGCAG[C/T]CTCATGTTGTCCTGTCTCAGGTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
55 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611609 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SIPA1L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SIPA1L2 - signal induced proliferation associated 1 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020808.4 | 5547 | Silent Mutation | AGA,AGG | R,R 1695 | NP_065859.3 | |
XM_005273211.2 | 5547 | Silent Mutation | AGA,AGG | R,R 1695 | XP_005273268.1 | |
XM_005273212.4 | 5547 | Silent Mutation | AGA,AGG | R,R 1695 | XP_005273269.1 | |
XM_005273213.4 | 5547 | Silent Mutation | AGA,AGG | R,R 1695 | XP_005273270.1 | |
XM_011544243.2 | 5547 | Silent Mutation | AGA,AGG | R,R 1695 | XP_011542545.1 | |
XM_017001896.1 | 5547 | Silent Mutation | AGA,AGG | R,R 1695 | XP_016857385.1 | |
XM_017001897.1 | 5547 | Silent Mutation | AGA,AGG | R,R 1677 | XP_016857386.1 | |
XM_017001898.1 | 5547 | Silent Mutation | AGA,AGG | R,R 1677 | XP_016857387.1 |
Set Membership: |
HapMap |