Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAGCCCTGGGCACCTGCCTGCCCT[C/T]CTCCTCTGCACCGCGGAAACTGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602052 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GAK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GAK - cyclin G associated kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318134.1 | Intron | NP_001305063.1 | ||||
NM_005255.3 | Intron | NP_005246.2 | ||||
XM_005272268.2 | Intron | XP_005272325.1 | ||||
XM_005272270.2 | Intron | XP_005272327.1 | ||||
XM_011513425.2 | Intron | XP_011511727.1 | ||||
XM_011513426.2 | Intron | XP_011511728.1 | ||||
XM_011513427.2 | Intron | XP_011511729.1 | ||||
XM_011513428.2 | Intron | XP_011511730.1 | ||||
XM_011513429.2 | Intron | XP_011511731.1 | ||||
XM_011513430.1 | Intron | XP_011511732.1 | ||||
XM_011513431.2 | Intron | XP_011511733.1 | ||||
XM_011513432.2 | Intron | XP_011511734.1 | ||||
XM_011513434.2 | Intron | XP_011511736.1 | ||||
XM_017007991.1 | Intron | XP_016863480.1 | ||||
XM_017007992.1 | Intron | XP_016863481.1 | ||||
XM_017007993.1 | Intron | XP_016863482.1 | ||||
XM_017007994.1 | Intron | XP_016863483.1 | ||||
XM_017007995.1 | Intron | XP_016863484.1 |