Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCACACCTCTCTCTTCGCTACCG[C/T]GTTACCTAAAACAAATCAAAATAAG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
FAM19A4 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
FAM19A4 - family with sequence similarity 19 member A4, C-C motif chemokine like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005527.2 | 745 | Silent Mutation | ACA,ACG | T,T 139 | NP_001005527.1 | |
NM_182522.4 | 745 | Silent Mutation | ACA,ACG | T,T 139 | NP_872328.1 | |
XM_011533371.1 | 745 | Missense Mutation | ACG,GCG | T,A 126 | XP_011531673.1 | |
XM_011533372.1 | 745 | Missense Mutation | ACG,GCG | T,A 126 | XP_011531674.1 |