Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCACCCAGGCCGGGAGCTCCAGG[C/G]TACCAGCTCCCTTCCTGTCTCCGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605350 | ||||||||||||||||||||
Literature Links: |
IL27RA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IL27RA - interleukin 27 receptor subunit alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PALM3 - paralemmin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145028.1 | 779 | Missense Mutation | ACC,AGC | T,S 260 | NP_001138500.1 | |
XM_005259891.4 | 779 | Missense Mutation | ACC,AGC | T,S 275 | XP_005259948.1 | |
XM_011527965.2 | 779 | Missense Mutation | ACC,AGC | T,S 248 | XP_011526267.1 |