Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600734 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C11orf45 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
C11orf45 - chromosome 11 open reading frame 45 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256088.1 | 2726 | UTR 3 | NP_001243017.1 | |||
NM_145013.2 | 2726 | UTR 3 | NP_659450.1 | |||
XM_011542654.2 | 2726 | UTR 3 | XP_011540956.1 | |||
XM_011542655.2 | 2726 | UTR 3 | XP_011540957.1 |
KCNJ5 - potassium voltage-gated channel subfamily J member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000890.3 | 2726 | Intron | NP_000881.3 | |||
XM_011542809.2 | 2726 | Intron | XP_011541111.1 | |||
XM_011542810.2 | 2726 | Intron | XP_011541112.1 |
Set Membership: |
HapMap Validated |