Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604742 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCLK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
DCLK1 - doublecortin like kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195415.1 | 4170 | UTR 3 | NP_001182344.1 | |||
NM_001195416.1 | 4170 | UTR 3 | NP_001182345.1 | |||
NM_001195430.1 | 4170 | Intron | NP_001182359.1 | |||
NM_004734.4 | 4170 | UTR 3 | NP_004725.1 | |||
XM_005266592.2 | 4170 | UTR 3 | XP_005266649.1 | |||
XM_006719893.2 | 4170 | UTR 3 | XP_006719956.1 | |||
XM_017020847.1 | 4170 | UTR 3 | XP_016876336.1 | |||
XM_017020848.1 | 4170 | Intron | XP_016876337.1 |
Set Membership: |
HapMap Validated |