Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615210 MIM: 615423 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCNP PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
PCNP - PEST proteolytic signal containing nuclear protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320395.1 | Intron | NP_001307324.1 | ||||
NM_001320397.1 | Intron | NP_001307326.1 | ||||
NM_001320398.1 | Intron | NP_001307327.1 | ||||
NM_001320399.1 | Intron | NP_001307328.1 | ||||
NM_001320400.1 | Intron | NP_001307329.1 | ||||
NM_001320401.1 | Intron | NP_001307330.1 | ||||
NM_020357.2 | Intron | NP_065090.1 | ||||
XM_017006922.1 | Intron | XP_016862411.1 |
TRMT10C - tRNA methyltransferase 10C, mitochondrial RNase P subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |