Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606738 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OSBPL10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
OSBPL10 - oxysterol binding protein like 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001174060.1 | 2293 | UTR 3 | NP_001167531.1 | |||
NM_017784.4 | 2293 | UTR 3 | NP_060254.2 | |||
XM_005264843.3 | 2293 | Intron | XP_005264900.1 | |||
XM_005264844.4 | 2293 | Intron | XP_005264901.1 | |||
XM_005264845.1 | 2293 | UTR 3 | XP_005264902.1 | |||
XM_011533326.2 | 2293 | Intron | XP_011531628.1 | |||
XM_017005667.1 | 2293 | Intron | XP_016861156.1 | |||
XM_017005668.1 | 2293 | Intron | XP_016861157.1 | |||
XM_017005669.1 | 2293 | Intron | XP_016861158.1 | |||
XM_017005670.1 | 2293 | Intron | XP_016861159.1 |