Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGTGTCATTACAGCAGCTGACAT[A/G]TGGAAATGTCAGTCCAATACAGACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614121 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAPD4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PAPD4 - PAP associated domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114393.1 | 1017 | Missense Mutation | TAT,TGT | Y,C 58 | NP_001107865.1 | |
NM_001114394.1 | 1017 | Missense Mutation | TAT,TGT | Y,C 58 | NP_001107866.1 | |
NM_001297744.1 | 1017 | Missense Mutation | TAT,TGT | Y,C 58 | NP_001284673.1 | |
NM_001297745.1 | 1017 | Missense Mutation | TAT,TGT | Y,C 58 | NP_001284674.1 | |
NM_173797.3 | 1017 | Missense Mutation | TAT,TGT | Y,C 58 | NP_776158.2 | |
XM_011543219.2 | 1017 | Missense Mutation | TAT,TGT | Y,C 58 | XP_011541521.1 | |
XM_011543220.2 | 1017 | Missense Mutation | TAT,TGT | Y,C 58 | XP_011541522.1 | |
XM_011543221.2 | 1017 | Missense Mutation | TAT,TGT | Y,C 58 | XP_011541523.1 | |
XM_011543222.2 | 1017 | Missense Mutation | TAT,TGT | Y,C 58 | XP_011541524.1 | |
XM_011543223.2 | 1017 | Missense Mutation | TAT,TGT | Y,C 58 | XP_011541525.1 | |
XM_011543224.2 | 1017 | Missense Mutation | TAT,TGT | Y,C 58 | XP_011541526.1 | |
XM_011543225.2 | 1017 | Missense Mutation | TAT,TGT | Y,C 58 | XP_011541527.1 | |
XM_011543226.2 | 1017 | Missense Mutation | TAT,TGT | Y,C 58 | XP_011541528.1 | |
XM_011543227.2 | 1017 | Missense Mutation | TAT,TGT | Y,C 58 | XP_011541529.1 | |
XM_011543228.2 | 1017 | Missense Mutation | TAT,TGT | Y,C 58 | XP_011541530.1 | |
XM_011543229.2 | 1017 | Missense Mutation | TAT,TGT | Y,C 58 | XP_011541531.1 | |
XM_011543230.2 | 1017 | Missense Mutation | TAT,TGT | Y,C 58 | XP_011541532.1 | |
XM_011543231.2 | 1017 | Missense Mutation | TAT,TGT | Y,C 58 | XP_011541533.1 | |
XM_011543232.2 | 1017 | Intron | XP_011541534.1 | |||
XM_011543233.2 | 1017 | Intron | XP_011541535.1 | |||
XM_017009151.1 | 1017 | Missense Mutation | TAT,TGT | Y,C 58 | XP_016864640.1 | |
XM_017009152.1 | 1017 | Missense Mutation | TAT,TGT | Y,C 58 | XP_016864641.1 | |
XM_017009153.1 | 1017 | Missense Mutation | TAT,TGT | Y,C 58 | XP_016864642.1 | |
XM_017009154.1 | 1017 | Missense Mutation | TAT,TGT | Y,C 58 | XP_016864643.1 | |
XM_017009155.1 | 1017 | Missense Mutation | TAT,TGT | Y,C 58 | XP_016864644.1 | |
XM_017009156.1 | 1017 | Missense Mutation | TAT,TGT | Y,C 58 | XP_016864645.1 | |
XM_017009157.1 | 1017 | Missense Mutation | TAT,TGT | Y,C 58 | XP_016864646.1 | |
XM_017009158.1 | 1017 | Missense Mutation | TAT,TGT | Y,C 58 | XP_016864647.1 | |
XM_017009159.1 | 1017 | Missense Mutation | TAT,TGT | Y,C 58 | XP_016864648.1 | |
XM_017009160.1 | 1017 | Missense Mutation | TAT,TGT | Y,C 58 | XP_016864649.1 | |
XM_017009161.1 | 1017 | UTR 5 | XP_016864650.1 | |||
XM_017009162.1 | 1017 | UTR 5 | XP_016864651.1 |