Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGAGACAGACAGACGCACGCACA[C/G]GCACACACGCAGCCATGCCGCTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300680 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PNCK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PNCK - pregnancy up-regulated nonubiquitous CaM kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039582.3 | 746 | Intron | NP_001034671.3 | |||
NM_001135740.1 | 746 | Intron | NP_001129212.1 | |||
XM_005274651.2 | 746 | Intron | XP_005274708.1 | |||
XM_006724809.3 | 746 | Intron | XP_006724872.1 | |||
XM_011531107.2 | 746 | Intron | XP_011529409.2 | |||
XM_011531108.2 | 746 | Intron | XP_011529410.1 | |||
XM_011531110.2 | 746 | Intron | XP_011529412.1 | |||
XM_011531111.2 | 746 | Intron | XP_011529413.1 | |||
XM_011531112.2 | 746 | Intron | XP_011529414.1 | |||
XM_017029277.1 | 746 | Intron | XP_016884766.1 | |||
XM_017029278.1 | 746 | Intron | XP_016884767.1 | |||
XM_017029279.1 | 746 | Intron | XP_016884768.1 | |||
XM_017029280.1 | 746 | UTR 3 | XP_016884769.1 |