Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACCGCGGTGAGAAACACAGCCGA[C/T]GCCTCACTCAGCGGCCGTTTCCCCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605796 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC186 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC186 - coiled-coil domain containing 186 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321829.1 | 167 | UTR 5 | NP_001308758.1 | |||
NM_018017.3 | 167 | UTR 5 | NP_060487.2 | |||
NM_153249.1 | 167 | UTR 5 | NP_694981.1 | |||
XM_011539915.2 | 167 | UTR 5 | XP_011538217.1 | |||
XM_011539916.2 | 167 | UTR 5 | XP_011538218.1 |
MIR2110 - microRNA 2110 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TDRD1 - tudor domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198795.1 | 167 | Intron | NP_942090.1 | |||
XM_005269978.2 | 167 | Intron | XP_005270035.1 | |||
XM_005269979.2 | 167 | Intron | XP_005270036.1 | |||
XM_011539959.2 | 167 | Intron | XP_011538261.1 | |||
XM_011539960.2 | 167 | Intron | XP_011538262.1 | |||
XM_011539961.2 | 167 | Intron | XP_011538263.1 | |||
XM_011539962.1 | 167 | Intron | XP_011538264.1 | |||
XM_011539963.1 | 167 | Intron | XP_011538265.1 | |||
XM_011539964.1 | 167 | Intron | XP_011538266.1 | |||
XM_011539966.1 | 167 | Intron | XP_011538268.1 | |||
XM_017016414.1 | 167 | Intron | XP_016871903.1 | |||
XM_017016415.1 | 167 | Intron | XP_016871904.1 |
Set Membership: |
HapMap |