Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTGGCTGCTGCTTTGAGGATGTT[G/T]CATACTTCAGGTCGGTGGAAATCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614109 MIM: 613901 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BPIFC PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BPIFC - BPI fold containing family C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_174932.2 | 1573 | Missense Mutation | GAA,GCA | E,A 479 | NP_777592.1 | |
XM_011530088.2 | 1573 | Missense Mutation | GAA,GCA | E,A 479 | XP_011528390.1 | |
XM_011530089.1 | 1573 | Missense Mutation | GAA,GCA | E,A 479 | XP_011528391.1 | |
XM_011530090.1 | 1573 | Missense Mutation | GAA,GCA | E,A 479 | XP_011528392.1 | |
XM_011530091.2 | 1573 | Missense Mutation | GAA,GCA | E,A 427 | XP_011528393.1 | |
XM_017028740.1 | 1573 | Missense Mutation | GAA,GCA | E,A 422 | XP_016884229.1 |
RTCB - RNA 2',3'-cyclic phosphate and 5'-OH ligase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |