Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACCTGCTGTAACAATGCTGACTG[C/T]GCAGCCGCATTATGCTGTAATTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PROSER1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PROSER1 - proline and serine rich 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025138.4 | 3498 | Silent Mutation | GCA,GCG | A,A 897 | NP_079414.3 | |
XM_005266545.3 | 3498 | Silent Mutation | GCA,GCG | A,A 875 | XP_005266602.1 | |
XM_011535239.2 | 3498 | Silent Mutation | GCA,GCG | A,A 868 | XP_011533541.1 |