Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTAAGCTGGTGACGTGCTGGACT[C/T]ACACTTGGGGGTTCCACACTTGTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRP12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRP12 - LDL receptor related protein 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135703.2 | 2284 | Silent Mutation | GTA,GTG | V,V 700 | NP_001129175.1 | |
NM_013437.4 | 2284 | Silent Mutation | GTA,GTG | V,V 719 | NP_038465.1 |
MIR548A3 - microRNA 548a-3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |