Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTATTTAACACAACTAAAAAATC[C/T]GAACAAAACAGCCTTATTTTTCCTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600620 MIM: 616234 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FKBP1B PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FKBP1B - FK506 binding protein 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322963.1 | Intron | NP_001309892.1 | ||||
NM_001322964.1 | Intron | NP_001309893.1 | ||||
NM_004116.4 | Intron | NP_004107.1 | ||||
NM_054033.3 | Intron | NP_473374.1 | ||||
XM_011532699.2 | Intron | XP_011531001.1 | ||||
XM_017003593.1 | Intron | XP_016859082.1 | ||||
XM_017003594.1 | Intron | XP_016859083.1 | ||||
XM_017003595.1 | Intron | XP_016859084.1 | ||||
XM_017003596.1 | Intron | XP_016859085.1 |
MFSD2B - major facilitator superfamily domain containing 2B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDCP - WD repeat and coiled coil containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142319.1 | Intron | NP_001135791.1 | ||||
NM_025203.2 | Intron | NP_079479.1 | ||||
XM_017005029.1 | Intron | XP_016860518.1 |
Set Membership: |
HapMap |