Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEC14L6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SEC14L6 - SEC14 like lipid binding 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193336.2 | Intron | NP_001180265.2 | ||||
XM_011530357.2 | Intron | XP_011528659.1 | ||||
XM_011530358.2 | Intron | XP_011528660.1 | ||||
XM_011530360.2 | Intron | XP_011528662.1 | ||||
XM_017028931.1 | Intron | XP_016884420.1 | ||||
XM_017028932.1 | Intron | XP_016884421.1 | ||||
XM_017028933.1 | Intron | XP_016884422.1 | ||||
XM_017028934.1 | Intron | XP_016884423.1 |