Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGTTTAACAAATCAGGAGCCTAT[A/T]GCTGTGGAGAATGTTGAAGCTTTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTBD8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BTBD8 - BTB domain containing 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_183242.3 | 570 | Silent Mutation | ATA,ATT | I,I 101 | NP_899065.2 |