Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGCCTCCCACCCACAGGCTTCGCA[A/G]GGCATGGTGGGTCAGCTGGCGGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604788 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6798 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR6798 - microRNA 6798 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RUVBL2 - RuvB like AAA ATPase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321190.1 | 666 | Silent Mutation | CAA,CAG | Q,Q 10 | NP_001308119.1 | |
NM_001321191.1 | 666 | UTR 5 | NP_001308120.1 | |||
NM_006666.2 | 666 | Silent Mutation | CAA,CAG | Q,Q 44 | NP_006657.1 | |
XM_011526330.1 | 666 | UTR 5 | XP_011524632.1 |