Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATAGAAGTGCTGAGGCGCATCCC[A/G]GAGCCGCATCTCCATCAGTGGCTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614730 MIM: 608292 | ||||||||||||||||||||
Literature Links: |
PIGO PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PIGO - phosphatidylinositol glycan anchor biosynthesis class O | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201484.1 | 2238 | Missense Mutation | CGG,TGG | R,W 582 | NP_001188413.1 | |
NM_032634.3 | 2238 | Missense Mutation | CGG,TGG | R,W 999 | NP_116023.2 | |
NM_152850.3 | 2238 | Missense Mutation | CGG,TGG | R,W 582 | NP_690577.2 | |
XM_005251619.3 | 2238 | Missense Mutation | CGG,TGG | R,W 999 | XP_005251676.1 | |
XM_017015222.1 | 2238 | Missense Mutation | CGG,TGG | R,W 999 | XP_016870711.1 | |
XM_017015223.1 | 2238 | Missense Mutation | CGG,TGG | R,W 582 | XP_016870712.1 | |
XM_017015224.1 | 2238 | Missense Mutation | CGG,TGG | R,W 582 | XP_016870713.1 |
STOML2 - stomatin like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |