Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGTGAAGCAGCTGTTAATTAGA[G/T]GAGGCCTTTGTCTGTAAGGAACCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKDD1B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ANKDD1B - ankyrin repeat and death domain containing 1B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POC5 - POC5 centriolar protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099271.1 | Intron | NP_001092741.1 | ||||
NM_152408.2 | Intron | NP_689621.2 | ||||
XM_005248436.1 | Intron | XP_005248493.1 | ||||
XM_011543158.2 | Intron | XP_011541460.1 | ||||
XM_011543159.1 | Intron | XP_011541461.1 | ||||
XM_011543160.1 | Intron | XP_011541462.1 | ||||
XM_017009037.1 | Intron | XP_016864526.1 | ||||
XM_017009038.1 | Intron | XP_016864527.1 | ||||
XM_017009039.1 | Intron | XP_016864528.1 | ||||
XM_017009040.1 | Intron | XP_016864529.1 |