Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGAGAATCACACCACAGAGACCA[A/G]TCTTCACAAAAAGGGTCCAACATTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609690 MIM: 612827 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FARSB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FARSB - phenylalanyl-tRNA synthetase beta subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005687.4 | 2056 | UTR 3 | NP_005678.3 | |||
XM_006712169.2 | 2056 | UTR 3 | XP_006712232.1 | |||
XM_011510466.2 | 2056 | UTR 3 | XP_011508768.1 | |||
XM_017003110.1 | 2056 | UTR 3 | XP_016858599.1 |
SGPP2 - sphingosine-1-phosphate phosphatase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |