Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGCACACAACGACTGGTTGTCAT[C/T]TTGTGGGACTGGTCTCCATTATTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612833 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DHRS7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DHRS7 - dehydrogenase/reductase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322280.1 | 1081 | Silent Mutation | AAA,AAG | K,K 213 | NP_001309209.1 | |
NM_001322281.1 | 1081 | Silent Mutation | AAA,AAG | K,K 123 | NP_001309210.1 | |
NM_001322282.1 | 1081 | Silent Mutation | AAA,AAG | K,K 183 | NP_001309211.1 | |
NM_016029.3 | 1081 | Silent Mutation | AAA,AAG | K,K 263 | NP_057113.1 | |
XM_017021368.1 | 1081 | Silent Mutation | AAA,AAG | K,K 263 | XP_016876857.1 |
PCNX4 - pecanex homolog 4 (Drosophila) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |