Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180630 MIM: 604983 | ||||||||||||||||||||||||||||||||
Literature Links: |
CEP95 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
CEP95 - centrosomal protein 95 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316990.1 | 22 | UTR 5 | NP_001303919.1 | |||
NM_138363.2 | 22 | UTR 5 | NP_612372.1 | |||
XM_005257779.3 | 22 | UTR 5 | XP_005257836.1 | |||
XM_005257780.2 | 22 | UTR 5 | XP_005257837.1 | |||
XM_006722166.3 | 22 | UTR 5 | XP_006722229.1 | |||
XM_011525458.2 | 22 | UTR 5 | XP_011523760.1 | |||
XM_017025287.1 | 22 | UTR 5 | XP_016880776.1 | |||
XM_017025288.1 | 22 | UTR 5 | XP_016880777.1 |
DDX5 - DEAD-box helicase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320595.1 | 22 | UTR 5 | NP_001307524.1 | |||
NM_001320596.1 | 22 | Intron | NP_001307525.1 | |||
NM_001320597.1 | 22 | Intron | NP_001307526.1 | |||
NM_004396.4 | 22 | Intron | NP_004387.1 |
MIR3064 - microRNA 3064 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR5047 - microRNA 5047 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLG2 - DNA polymerase gamma 2, accessory subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |