Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGACAGGTGGGTTCTGAGGCGGT[A/G]GGTCCTCCGAAAGGAAGGGGCTTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182175 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SRP19 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SRP19 - signal recognition particle 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204193.1 | Intron | NP_001191122.1 | ||||
NM_001204194.1 | Intron | NP_001191123.1 | ||||
NM_001204196.1 | Intron | NP_001191125.1 | ||||
NM_001204199.1 | Intron | NP_001191128.1 | ||||
NM_003135.2 | Intron | NP_003126.1 |