Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCCACAAAGGACACCCAAATCCT[A/T]GGGATATGGCACATTAAAAGATCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
INO80C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
INO80C - INO80 complex subunit C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098817.1 | Intron | NP_001092287.1 | ||||
NM_001308064.1 | Intron | NP_001294993.1 | ||||
NM_194281.3 | Intron | NP_919257.2 | ||||
XM_005258196.4 | Intron | XP_005258253.1 | ||||
XM_005258197.4 | Intron | XP_005258254.1 | ||||
XM_011525812.2 | Intron | XP_011524114.1 | ||||
XM_017025549.1 | Intron | XP_016881038.1 |