Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTACCAATGTGACTCTGTCCTCTG[A/G]CTTTGTGGCTGACAGAGGTGTAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602486 MIM: 602487 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
POP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
POP1 - POP1 homolog, ribonuclease P/MRP subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145860.1 | 102 | Missense Mutation | GAC,GGC | D,G 25 | NP_001139332.1 | |
NM_001145861.1 | 102 | Missense Mutation | GAC,GGC | D,G 25 | NP_001139333.1 | |
NM_015029.2 | 102 | Missense Mutation | GAC,GGC | D,G 25 | NP_055844.2 | |
XM_011516801.2 | 102 | Missense Mutation | GAC,GGC | D,G 25 | XP_011515103.1 |
RIDA - reactive intermediate imine deaminase A homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |