Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGAGCCGCGTCGGCGGCCACGGC[A/G]GCGCCCACGGCCACTCCCGCCGCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603051 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGPS PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AGPS - alkylglycerone phosphate synthase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003659.3 | 254 | Silent Mutation | GCA,GCG | A,A 69 | NP_003650.1 | |
XM_011512041.2 | 254 | Intron | XP_011510343.1 | |||
XM_011512042.1 | 254 | Intron | XP_011510344.1 | |||
XM_011512043.2 | 254 | Intron | XP_011510345.1 |
LOC100130691 - uncharacterized LOC100130691 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |