Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCACAGGGAATACCTTTGTCAC[C/G]AGCACCAAACTGTGAAGGCACTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608918 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATPAF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATPAF2 - ATP synthase mitochondrial F1 complex assembly factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GID4 - GID complex subunit 4 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024052.4 | Intron | NP_076957.3 | ||||
XM_005256799.4 | Intron | XP_005256856.1 | ||||
XM_005256800.1 | Intron | XP_005256857.1 | ||||
XM_011524008.1 | Intron | XP_011522310.1 |