Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGTTCGGTAGTCCTGAGGAGGGT[A/G]AGGGCTGTGTGTGTGGTCTTGTGGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608305 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC13A5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SLC13A5 - solute carrier family 13 member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143838.2 | 1697 | UTR 3 | NP_001137310.1 | |||
NM_001284509.1 | 1697 | UTR 3 | NP_001271438.1 | |||
NM_001284510.1 | 1697 | UTR 3 | NP_001271439.1 | |||
NM_177550.4 | 1697 | UTR 3 | NP_808218.1 | |||
XM_011523795.2 | 1697 | Intron | XP_011522097.1 |
Set Membership: |
HapMap |