Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCTCTGCGGCAGGTGGAGGCGGC[A/C]CGCAAGGCCATCCGCGCCGCTCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC84 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC84 - coiled-coil domain containing 84 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198489.2 | 197 | Silent Mutation | GCA,GCC | A,A 44 | NP_940891.1 | |
XM_011542799.1 | 197 | UTR 5 | XP_011541101.1 | |||
XM_011542800.2 | 197 | Intron | XP_011541102.1 | |||
XM_017017652.1 | 197 | UTR 5 | XP_016873141.1 |
RPL23AP64 - ribosomal protein L23a pseudogene 64 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |