Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606236 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASPSCR1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ASPSCR1 - ASPSCR1, UBX domain containing tether for SLC2A4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001251888.1 | 474 | Silent Mutation | CCA,CCG | P,P 122 | NP_001238817.1 | |
NM_024083.3 | 474 | Silent Mutation | CCA,CCG | P,P 122 | NP_076988.1 | |
XM_011523601.2 | 474 | Silent Mutation | CCA,CCG | P,P 122 | XP_011521903.1 | |
XM_011523602.2 | 474 | Silent Mutation | CCA,CCG | P,P 122 | XP_011521904.1 | |
XM_017025036.1 | 474 | Silent Mutation | CCA,CCG | P,P 122 | XP_016880525.1 | |
XM_017025037.1 | 474 | Silent Mutation | CCA,CCG | P,P 122 | XP_016880526.1 | |
XM_017025038.1 | 474 | Silent Mutation | CCA,CCG | P,P 45 | XP_016880527.1 | |
XM_017025039.1 | 474 | Silent Mutation | CCA,CCG | P,P 45 | XP_016880528.1 | |
XM_017025040.1 | 474 | Silent Mutation | CCA,CCG | P,P 45 | XP_016880529.1 | |
XM_017025041.1 | 474 | Silent Mutation | CCA,CCG | P,P 45 | XP_016880530.1 | |
XM_017025042.1 | 474 | Silent Mutation | CCA,CCG | P,P 122 | XP_016880531.1 | |
XM_017025043.1 | 474 | Silent Mutation | CCA,CCG | P,P 45 | XP_016880532.1 | |
XM_017025044.1 | 474 | Silent Mutation | CCA,CCG | P,P 45 | XP_016880533.1 |