Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF501 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZNF501 - zinc finger protein 501 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF502 - zinc finger protein 502 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134440.1 | 1580 | Missense Mutation | CCG,CTG | P,L 28 | NP_001127912.1 | |
NM_001134441.1 | 1580 | Missense Mutation | CCG,CTG | P,L 28 | NP_001127913.1 | |
NM_001134442.1 | 1580 | Missense Mutation | CCG,CTG | P,L 28 | NP_001127914.1 | |
NM_001282880.1 | 1580 | Missense Mutation | CCG,CTG | P,L 28 | NP_001269809.1 | |
NM_033210.4 | 1580 | Missense Mutation | CCG,CTG | P,L 28 | NP_149987.2 | |
XM_017007440.1 | 1580 | Missense Mutation | CCG,CTG | P,L 28 | XP_016862929.1 |
Set Membership: |
HapMap |