Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATTACAGGAAAGTAATTTCAAAA[A/T]TAGAATTTTTTTTTTTACCATAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616527 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFAP43 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CFAP43 - cilia and flagella associated protein 43 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SFR1 - SWI5 dependent homologous recombination repair protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002759.1 | Intron | NP_001002759.1 | ||||
NM_145247.4 | Intron | NP_660290.3 | ||||
XM_005269521.3 | Intron | XP_005269578.1 | ||||
XM_017015672.1 | Intron | XP_016871161.1 |