Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTTTTGCCTCTCTCTTCTGAACT[A/G]TCTGGGGTAACACATTATTTTGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612486 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCHS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCHS2 - dachsous cadherin-related 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142552.1 | 5486 | Intron | NP_001136024.1 | |||
NM_017639.3 | 5486 | Missense Mutation | ACA,ATA | T,I 2731 | NP_060109.2 | |
XM_011532045.1 | 5486 | Missense Mutation | ACA,ATA | T,I 3186 | XP_011530347.1 | |
XM_011532046.2 | 5486 | Missense Mutation | ACA,ATA | T,I 2409 | XP_011530348.1 | |
XM_011532049.2 | 5486 | Missense Mutation | ACA,ATA | T,I 1521 | XP_011530351.1 | |
XM_017008320.1 | 5486 | Intron | XP_016863809.1 |