Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGATGATGAGCAGCAGCAGGCC[A/G]GGCTGCAGGGGAGAGAAGCCGAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610875 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6804 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR6804 - microRNA 6804 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP6R1 - protein phosphatase 6 regulatory subunit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM86B - transmembrane protein 86B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173804.4 | 662 | Silent Mutation | CCC,CCT | P,P 122 | NP_776165.3 | |
XM_017026554.1 | 662 | Silent Mutation | CCC,CCT | P,P 121 | XP_016882043.1 |