Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCAAAGACAGGTGGGCTCTGTCC[C/T]GCCTTCACCTGGAGCTGCCCTGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601481 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYO10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYO10 - myosin X | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012334.2 | 5544 | UTR 3 | NP_036466.2 | |||
XM_005248306.4 | 5544 | UTR 3 | XP_005248363.1 | |||
XM_005248307.2 | 5544 | UTR 3 | XP_005248364.1 | |||
XM_006714475.2 | 5544 | UTR 3 | XP_006714538.1 | |||
XM_011514046.2 | 5544 | UTR 3 | XP_011512348.1 |