Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602034 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRHR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRHR2 - corticotropin releasing hormone receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202475.1 | Intron | NP_001189404.1 | ||||
NM_001202481.1 | Intron | NP_001189410.1 | ||||
NM_001202482.1 | Intron | NP_001189411.1 | ||||
NM_001202483.1 | Intron | NP_001189412.1 | ||||
NM_001883.4 | Intron | NP_001874.2 | ||||
XM_011515127.2 | Intron | XP_011513429.1 | ||||
XM_011515128.2 | Intron | XP_011513430.1 | ||||
XM_011515129.2 | Intron | XP_011513431.1 | ||||
XM_017011751.1 | Intron | XP_016867240.1 | ||||
XM_017011752.1 | Intron | XP_016867241.1 |