Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTTCAGCACCTGGTGCAGGTGCC[A/G]CATCTCCTGCCGGTACTGGGCTGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615470 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP89 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CEP89 - centrosomal protein 89 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032816.4 | 2226 | Silent Mutation | NP_116205.3 | |||
XM_005259344.3 | 2226 | Missense Mutation | XP_005259401.1 | |||
XM_011527425.2 | 2226 | Missense Mutation | XP_011525727.1 | |||
XM_017027398.1 | 2226 | Intron | XP_016882887.1 |